(±)-2-Phenylpiperazine, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
638641-25G
638641-5G
SIAL638641-25GEA
17260
CZK
SIAL638641-25G
SIAL638641-5G
(±)-2-Phenylpiperazine, Sigma-Aldrich®
(±)-2-Phenylpiperazine
Vzorec:
C₁₀H₁₄N₂ MW: 162,23 g/mol |
MDL Number:
MFCD01871362 CAS číslo: 5271-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...