Tisk…

(±)-trans-1,2-Cyclopentanediol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-trans-Cyclopentane-1,2-diol, trans-Cyclopentane-1,2-diol

M-Clarity™ quality level = MQ200

361445-5G 361445-1G
SIAL361445-5GEA 6430 CZK
SIAL361445-5G SIAL361445-1G
(±)-trans-1,2-Cyclopentanediol, Sigma-Aldrich®
(±)-trans-1,2-Cyclopentanediol
Vzorec: C₅H₁₀O₂
MW: 102,13 g/mol
Bod varu: 229 °C (1013 hPa)
Bod tání: 54…49 °C
MDL Number: MFCD00082582
CAS číslo: 5057-99-8
EINECS: 225-757-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR