Tisk…

(±)-1-Phenyl-1,2-ethanediol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-α,β-Dihydroxyethylbenzene, (±)-Phenylethane-1,2-diol, 1-Phenylethane-1,2-diol

M-Clarity™ quality level = MQ100

P24055-100G P24055-5G
SIALP24055-100GEA 9350 CZK
SIALP24055-100G SIALP24055-5G
(±)-1-Phenyl-1,2-ethanediol, Sigma-Aldrich®
(±)-1-Phenyl-1,2-ethanediol
Vzorec: C₆H₅CH(OH)CH₂OH
MW: 138,17 g/mol
Bod varu: 272…274 °C (1013 hPa)
Bod tání: 61…64 °C
MDL Number: MFCD00003546
CAS číslo: 93-56-1
EINECS: 202-258-1
Merck Index: 12,09029

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR