Tisk…

3-Chloro-α-methylbenzyl alcohol ≥97%

Dodavatel: Thermo Scientific

Synonyms: 3-Chlorophenyl methyl carbinol, 1-(3-Chlorophenyl)ethanol, m-Chloro-α-methylbenzyl alcohol

L04783.06 L04783.14
L04783.06EA 1480 CZK
L04783.06 L04783.14
3-Chloro-α-methylbenzyl alcohol ≥97%
3-Chloro-α-methylbenzyl alcohol

Purity: 97% CAS No.: 6939-95-3 Molecular Formula: C8H9ClO Formula Weight: 156.61 Flash Point: 110°(230°F) Refractive Index: 1.5445 Boiling Point: 83-85°/1.5mm Beilstein Reference: 2042021 MDL No.: MFCD00021864

Vzorec: C₈H₉ClO
MW: 156,61 g/mol
Bod varu: 83…85 °C (1,5 mmHg)
Hustota: 1,173
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00021864
CAS číslo: 6939-95-3
EINECS: 230-079-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR