Tisk…

(±)-4-(Trifluoromethyl)mandelic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (±)-4-(Trifluoromethyl)phenylglycolic acid, 2-Hydroxy-2-(4-(trifluoromethyl)phenyl)acetic acid

B22264.03 B22264.06
B22264.03EA 948 CZK
B22264.03 B22264.06
(±)-4-(Trifluoromethyl)mandelic acid ≥98%
(±)-4-(Trifluoromethyl)mandelic acid

Purity: 98% CAS No.: 395-35-7 Molecular Formula: C9H7F3O3 Formula Weight: 220.15 Melting Point: 129-131° Beilstein Reference: 2979949

Vzorec: C₉H₇F₃O₃
MW: 220.15
Bod varu: 314
Bod tání: 129…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00009724
CAS číslo: 395-35-7
EINECS: 206-900-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR