Tisk…

(±)-Mandelic acid, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid

M-Clarity™ quality level = MQ200

M2101-250G M2101-500G
SIALM2101-250GEA 2370 CZK
SIALM2101-250G SIALM2101-500G
(±)-Mandelic acid, Sigma-Aldrich®
(±)-Mandelic acid
Vzorec: C₈H₈O₃
Bod varu: 322 °C (1013 hPa)
Bod tání: 117…120 °C
Hustota: 1,361 g/cm³ (20 °C)
MDL Number: MFCD00064250
CAS číslo: 90-64-2
EINECS: 210-277-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR