(±)-4-Chloromandelic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Chloromandelic acid, (±)-4-Chlorophenylglycolic acid
B24264.06
B24264.14
B24264.06EA
1340
CZK
B24264.06
B24264.14
(±)-4-Chloromandelic acid ≥98%
(±)-4-Chloromandelic acid
Purity: 98%
CAS No.: 492-86-4
Molecular Formula: C8H7ClO3
Formula Weight: 186.60
Melting Point: 117-122°
Beilstein Reference: 1567759
Vzorec:
C₈H₇ClO₃ Bod varu: 361 °C (1013 hPa) Bod tání: 117…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042724 CAS číslo: 492-86-4 EINECS: 207-764-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...