Tisk…

Saponin, Sigma-Aldrich®

Dodavatel: Merck

84510-100G 84510-500G
SIAL84510-100GEA 3640 CZK
SIAL84510-100G SIAL84510-500G
Saponin, Sigma-Aldrich®
Saponin
Hustota: 1,015…1.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081981
CAS číslo: 8047-15-2
EINECS: 232-462-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR