Tisk…

Saponin (from Quillaja Bark), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

S7900-100G S7900-25G
SIALS7900-100GEA 8760 CZK
SIALS7900-100G SIALS7900-25G
Saponin (from Quillaja Bark), Sigma-Aldrich®
Saponin
MDL Number: MFCD00081981
CAS číslo: 8047-15-2
EINECS: 232-462-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR