6-Fluorescein dT phosphoramidite
Dodavatel: AAT Bioquest
Fluorescein-dT phosphoramidite provides a valuable tool to add the fluorescein tag to a desired location of an oligo sequence. For efficiently labeling an oligonucleotide.
- Spectral profile is compatible with all major fluorescence platforms
Fluorescein-dT phosphoramidite contains a DMT group to allow quantification of coupling. It can be inserted into a target sequence as a replacement for a dT residue.
It is complimentary to the commonly used 6-FAM phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis.
Balení: Plastic vial.
Upozornení: For research use only.
Vzorec:
C₇₉H₈₉N₆O₁₇P |
CAS číslo:
289712-99-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...