Tisk…

Biotin CE Phosphoramidite

Dodavatel: AAT BIOQUEST

Synonyms: N-[6-[[(Diisopropylamino)(2-cyanoethoxy)phosphino]oxy]-5-[(4,4'-dimethoxytrityloxy)methyl]hexyl]-5-(2-oxo-1,3,3abeta,4,6,6abeta-hexahydro-2H-thieno[3,4-d]imidazole-4alpha-yl)pentanamide, Biotin Phosphoramidite

6006 6222
AATB6006EA 65630 CZK
AATB6006 AATB6222
Biotin CE Phosphoramidite
Biotin CE Phosphoramidite

Biotin phosphoramidite is a valuable tool to add the biotin tag to an oligo sequence. For efficiently labeling an oligonucleotide.


  • DMT group allows coupling quantification, and can be used in RP cartridge and HPLC purification techniques
  • Thoroughly referenced


Biotin phosphoramidite contains a 15-atom spacer arm minimizing the potential hindrance of biotin binding.


It is complimentary to the commonly used biotin phosphoramidite that contains no DMT group and can only be added at the 5'-terminus, thus terminating synthesis.


Balení: Plastic vial.


Upozornení: For research use only.

Vzorec: C₄₇H₆₆N₅O₇PS
MW: 876,11 g/mol
CAS číslo: 147190-34-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR