Tisk…

Fmoc-Asp(Edans)-OH ≥90%

Dodavatel: AAT BIOQUEST
Varování

5000 5001 5002
AATB5000EA 4060 CZK
AATB5000 AATB5001 AATB5002
Fmoc-Asp(Edans)-OH ≥90%
Fmoc-Asp(Edans)-OH

FMOC-Asp(EDANS)-OH is a building block for in-sequence peptide Asp labeling by EDANS.

Vzorec: C₃₁H₂₉N₃O₈S
MDL Number: MFCD02094140
CAS číslo: 182253-73-2
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac5f20d3a345335247639abbfb5ce08925564831&vid=523a569de809d7174d42ff94a178b5a38b3cdce3&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR