Tisk…
California Red™ SE ≥90% Superior Replacement to Texas Red®
Dodavatel: AAT Bioquest
473 479
AATB473EA 6140 CZK
AATB473 AATB479
California Red™ SE ≥90% Superior Replacement to Texas Red®
California Red™ SE
Although Texas Red® is the most popular labeling reagent of sulfonyl chloride, it is quite unstable in water.
  • Identical spectra to Texas Red
  • High Stability
  • Improved Labeling Efficiency
Upozornení: For research use only.