Tisk…

6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) ≥90%

Dodavatel: AAT BIOQUEST
Varování

392 399
AATB392EA 4060 CZK
AATB392 AATB399
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE) ≥90%
6-Carboxy-X-rhodamine succinimidyl ester (6-ROX SE)

6-ROX, SE is the other purified single isomer of 5(6)-ROX, SE.

Vzorec: C₃₇H₃₃N₃O₇
CAS číslo: 216699-36-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3adeec2e618d117fc43365188bfd472486b11903&vid=240025677973921ee697127568bed3c40f01c362&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR