Tisk…

5-Carboxytetramethylrhodamine (5-TAMRA) ≥95%

Dodavatel: AAT BIOQUEST
Varování

Synonyms: 5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer

363 364
AATB363EA 3210 CZK
AATB363 AATB364
5-Carboxytetramethylrhodamine (5-TAMRA) ≥95%
5-Carboxytetramethylrhodamine (5-TAMRA)

5-TAMRA is the purified single isomer of 5(6)-TAMRA.

Vzorec: C₂₅H₂₂N₂O₅
MDL Number: MFCD00269769
CAS číslo: 91809-66-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a69abfcb05257a3d0e6e5d00246543a8c27ab383&vid=b096118283475b5bdd0437820922f5d2a524ed10&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR