Tisk…
6-VIC NHS ester (6-VIC SE) ≥90%
Dodavatel: AAT BIOQUEST
212 213
AATB212EA 7790 CZK
AATB212 AATB213
6-VIC NHS ester (6-VIC SE) ≥90%
6-VIC NHS ester (6-VIC SE)
6-VIC, SE is a popular amino-reactive fluorescent probe that is widely used in nucleic acid sequencing and related research.
  • Useful for Genetic Research
  • Excellent Quality
Upozornení: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfca7a3f984f7aca8330b8f95954ebc251f5969e&vid=3a0f7a62b9418b602533cfd11bd41b458c53d95f&prid=web&clid=SAPDAM