Tisk…

Coelenterazine h ≥95%

Dodavatel: AAT Bioquest
Varování

21153 21159 21165
AATB21153EA 3210 CZK
AATB21153 AATB21159 AATB21165
Coelenterazine h ≥95%
Coelenterazine h

Calcium measurement is critical for numerous biological investigations.

Vzorec: C₂₆H₂₁N₃O₂
MDL Number: MFCD00798394
CAS číslo: 50909-86-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38ed072e7d1aaa55a91b0450b4add4bbc11c425b&vid=5eed954b5a16e534f002fe5e8208158f1df26ae2&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR