Coelenterazine ≥95%
Dodavatel: AAT BIOQUEST
|
Varování
|
Synonyms:
CLZN
21150
21166
AATB21150EA
3210
CZK
AATB21150
AATB21166
Coelenterazine ≥95%
Coelenterazine
Calcium measurement is critical for numerous biological investigations.
Vzorec:
C₂₆H₂₁N₃O₃ MW: 423,472 g/mol |
MDL Number:
MFCD00467176 CAS číslo: 55779-48-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...