Fluo-8H™ AM
Dodavatel: AAT Bioquest
|
Varování
|
21090
21091
AATB21090EA
12280
CZK
AATB21090
AATB21091
Fluo-8H™ AM
Fluo-8H™ AM
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
Upozornení: For research use only.
ADR:
5,II |