Rhod-FF tripotassium salt
Dodavatel: AAT BIOQUEST
|
Varování
|
21075
21076
AATB21075EA
12280
CZK
AATB21075
AATB21076
Rhod-FF tripotassium salt
Rhod-FF tripotassium salt
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Low binding affinity
- Cell impermeant
Upozornení: For research use only.
MW:
891 g/mol |
ADR:
5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...