Tisk…
Calbryte™ 590 potassium salt
Dodavatel: AAT BIOQUEST
Varování
20706 20707
AATB20706EA 12280 CZK
AATB20706 AATB20707
Calbryte™ 590 potassium salt
Calbryte™ 590 potassium salt
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein & 'coupled receptors' (GPCRs) and calcium channel targets.
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell impermeant
Upozornení: For research use only.
ADR: 5,II