Tisk…

2',3'-cGAMP second messenger

Dodavatel: AAT Bioquest
Varování

20310 20311
AATB20310EA 18970 CZK
AATB20310 AATB20311
2',3'-cGAMP second messenger
2',3'-cGAMP

2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.


  • Useful for immunity research
  • Multiple applications

CAS číslo: 1441190-66-4
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8487368f2c6ff6b4a87432b6aa12879f1053ada&vid=a72b6153a87779407e422fec3806ebe083c5089f&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR