Tisk…
EDTA, AM ester metal ion chelator
Dodavatel: AAT BIOQUEST
Varování
19010 19011
AATB19010EA 4060 CZK
AATB19010 AATB19011
EDTA, AM ester metal ion chelator
EDTA, AM ester
Ethylenediaminetetraacetic acid (EDTA) might be the most popular metal ion chelator used for both industrial and medical purposes.
  • Excellent metal chelator
  • Multiple applications
MW: 580.5 g/mol
CAS číslo: 162303-59-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec4f685e9f6d8fcd7cae9a30a2126881928e46f&vid=6155ffacf55e8a1673beb43ba642717ddca623e5&prid=web&clid=SAPDAM