4-Methylumbelliferyl-β-D-galactopyranoside ≥95%
Dodavatel: AAT BIOQUEST
|
Varování
|
Synonyms:
4-Methylumbelliferyl β-D-galactopyranoside, 4-Methyl-7-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one
14010
14012
AATB14010EA
3210
CZK
AATB14010
AATB14012
4-Methylumbelliferyl-β-D-galactopyranoside ≥95%
4-Methylumbelliferyl-β-D-galactopyranoside
4-Methylumbelliferyl-beta-D-galactopyranoside (MUG) is a fluorogenic MU substrate for detecting galactosidase in cell extracts and purified enzyme preparations using a fluorescence microplate reader or fluorometer.
Vzorec:
C₁₆H₁₈O₈ MW: 338.312 g/mol |
MDL Number:
MFCD00036773 CAS číslo: 6160-78-7 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...