FAM-VAD-FMK ≥95%
Dodavatel: AAT BIOQUEST
13470
13484
AATB13470EA
4060
CZK
AATB13470
AATB13484
FAM-VAD-FMK ≥95%
FAM-VAD-FMK
FAM-VAD is a green fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
- Caspase Inhibitor
- High Intracellular Retention
- Useful In Multiple Cell Lines/Species
Upozornení: For research use only.