5-FAM-Alkyne ≥95%
Dodavatel: AAT Bioquest
132
951
AATB132EA
6140
CZK
AATB132
AATB951
5-FAM-Alkyne ≥95%
5-FAM-Alkyne
5-FAM alkyne is an excellent building block for developing fluoresceinated bioconjugates by reacting with an azide group.
- Multiple Bioconjugation Applications
- Excellent Quality
Upozornení: For research use only.
Vzorec:
C₂₄H₁₅NO₆ |
MDL Number:
MFCD28505571 CAS číslo: 510758-19-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...