Tisk…
Potassium deuteroxide 40% in D₂O, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
176761-10G 176761-50G
SIAL176761-10GEA 3240 CZK
SIAL176761-10G SIAL176761-50G
Potassium deuteroxide 40% in D₂O, Sigma-Aldrich®
Potassium deuteroxide
Vzorec: KOD
Storage Temperature: Ambient
CAS číslo: 24572-01-8
EINECS: 246-324-8
UN: 1814
ADR: 8,II