Tisk…

Anti-IgG Goat Polyclonal Antibody (iFluor® 568)

Dodavatel: AAT BIOQUEST INC
16692 16832
AATB16692EA 3900 CZK
AATB16692 AATB16832
Anti-IgG Goat Polyclonal Antibody (iFluor® 568)
Protilátky
iFluor® 568 is a bright red fluorescent dye.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Upozornení: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR