Tisk…
Stayright™ Purple, ReadiUse™ HRP Chromogen Premixed with Hydrogen Peroxide*
Dodavatel: AAT BIOQUEST
45900 45901
AATB45900EA 8190 CZK
AATB45900 AATB45901
Stayright™ Purple, ReadiUse™ HRP Chromogen Premixed with Hydrogen Peroxide*
Stayright™ Purple
3,3'-Diaminobenzidine (DAB) has been used as the most commonly used IHC chromogen because it is inexpensive and sensitive for routine applications.
  • Ready to use
  • Non-mutagenic
  • Minimal cytotoxicity
  • No mixing steps required
  • Stain persists through dehydration and coverslipping
Upozornení: For research use only.