Tisk…
Screen Quest™ TR-FRET No Wash cAMP Assay Kits
Dodavatel: AAT BIOQUEST
36380 36379 36381
AATB36380EA 35480 CZK
AATB36380 AATB36379 AATB36381
Screen Quest™ TR-FRET No Wash cAMP Assay Kits
Test Buněčná stanovení
Screen Quest™ TR-FRET No Wash cAMP Assay Kit provides a convenient assay method for monitoring the activation of adenylyl cyclase in G-protein coupled receptor systems.
  • Extremely sensitive detection
  • Contains all necessary components
  • No wash step required
  • Robust and adaptable protocol
  • Longer fluorescence lifetime
Upozornení: For research use only.