Chinhydron, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)
M-Clarity™ quality level = MQ100
282960-100G
282960-25G
SIAL282960-100GEA
4960
CZK
SIAL282960-100G
SIAL282960-25G
Chinhydron, Sigma-Aldrich®
Chinhydron
Vzorec:
C₆H₄(OH)₂·C₆H₄O₂ MW: 218,21 g/mol Bod tání: 168…171 °C Hustota: 1,35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010310 CAS číslo: 106-34-3 EINECS: 203-387-6 UN: 2811 ADR: 6.1,III Merck Index: 13,08148 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...