Hydrid sodný 60% dispersion in mineral oil, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
452912-5G
452912-100G
452912-2KG
452912-500G
452912-5KG
452912-1KG
SIAL452912-5GEA
1540
CZK
SIAL452912-5G
SIAL452912-100G
SIAL452912-2KG
SIAL452912-500G
SIAL452912-5KG
SIAL452912-1KG
Hydrid sodný 60% dispersion in mineral oil, Sigma-Aldrich®
Hydrid sodný
Vzorec:
HNa MW: 24 Storage Temperature: Ambient |
MDL Number:
MFCD00003471 CAS číslo: 7646-69-7 EINECS: 231-587-3 UN: 1427 ADR: 4.3 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...