Screen Quest™ Fluo-4 No Wash Calcium Assay Kit
Dodavatel: AAT BIOQUEST
36325
36326
AATB36325EA
16380
CZK
AATB36325
AATB36326
Screen Quest™ Fluo-4 No Wash Calcium Assay Kit
Test
Buněčná stanovení
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
- Well-cited calcium probe
- Contains all necessary components
- No wash step required
- Robust and adaptable protocol
- Multiple research applications
Upozornení: For research use only.