Tisk…
Screen Quest™ No Wash Potassium Channel Assay Kit
Dodavatel: AAT BIOQUEST
36550 36551 36552
AATB36550EA 8190 CZK
AATB36550 AATB36551 AATB36552
Screen Quest™ No Wash Potassium Channel Assay Kit
Test Buněčná stanovení
Potassium (K⁺) ion channel plays an important role in regulating fundamental biological processes including heart rate, hormone and neurotransmitter secretion, water and electrolyte balance.
  • Cell permeable potassium probe
  • No wash step required
  • Robust and adaptable protocol
  • Multiple research applications
Upozornení: For research use only.