Tisk…

Anti-IgG Goat Polyclonal Antibody [clone: IgG] (trFluor™ Eu)

Dodavatel: AAT BIOQUEST
16668 16820
AATB16668EA 12280 CZK
AATB16668 AATB16820
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (trFluor™ Eu)
Protilátky
Many biological compounds present in cells, serum or other biological fluids are naturally fluorescent, and thus the use of conventional, prompt fluorophores leads to serious limitations in assay sensitivity due to the high background caused by the autofluorescence of the biological molecules to be assayed.

  • Large stokes shift
  • Improved stability
  • High emission yield
  • Long emission half-life

Upozornení: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: trFluor™ Eu
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR