Fmoc-Glu(Edans)-OH ≥95%
Dodavatel: AAT BIOQUEST
|
Varování
|
5009
5010
5011
AATB5009EA
4060
CZK
AATB5009
AATB5010
AATB5011
Fmoc-Glu(Edans)-OH ≥95%
Fmoc-Glu(Edans)-OH
FMOC-Glu(EDANS)-OH is a building block for in-sequence peptide Glu labeling by EDANS.
Vzorec:
C₃₂H₃₁N₃O₈S MW: 617.673 g/mol |
MDL Number:
MFCD01076138 CAS číslo: 193475-66-0 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...