Tisk…
6-TAMRA azide ≥95%
Dodavatel: AAT BIOQUEST
490 965
AATB490EA 6140 CZK
AATB490 AATB965
6-TAMRA azide ≥95%
6-TAMRA azide
6-TAMRA aside is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Upozornení: For research use only.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b53386cf614f79129e452d1f9f6a98cf9c3f092&vid=6208ef18031e9ad2818e642cad3382b92224c726&prid=web&clid=SAPDAM