Tisk…

5-ROX ≥95% for molecular biology

Dodavatel: AAT BIOQUEST
Varování

385 381
AATB385EA 11720 CZK
AATB385 AATB381
5-ROX ≥95% for molecular biology
5-ROX

5-ROX is the purified single isomer of 5(6)-ROX.


  • Useful for multiple applications

Vzorec: C₃₃H₃₀N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD28355897
CAS číslo: 216699-35-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6234d68ecd61ce35c6414f160f819db1d4b8af8c&vid=279766db37d78d43ba9d26ff8a3f5d12dbba92c7&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR