6-TAMRA ≥95%
Dodavatel: AAT BIOQUEST
|
Varování
|
Synonyms:
6-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-4-carboxybenzoate
366
367
368
AATB366EA
3210
CZK
AATB366
AATB367
AATB368
6-TAMRA ≥95%
6-TAMRA
6-TAMRA is the purified single isomer of 5(6)-TAMRA.
Vzorec:
C₂₅H₂₂N₂O₅ |
MDL Number:
MFCD01073638 CAS číslo: 91809-67-5 ADR: 5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...