Tisk…
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) single isomer ≥90%
Dodavatel: AAT BIOQUEST
Synonyms: Carboxyrhodamine 6G succinimidyl ester, 6-isomer, 6-CR6G, NHS ester, 6-Carboxyrhodamine 6G, succinimidyl ester
342 346
AATB342EA 3210 CZK
AATB342 AATB346
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE) single isomer ≥90%
6-Carboxyrhodamine 6G succinimidyl ester (6-CR6G SE)
6-CR6G, SE is amine-reactive derivative of 6-CR6G.
  • Bright Fluorescence
  • High Photostability
  • Useful For Small Molecule Labeling
  • 90% Purity
Upozornení: For research use only.
Vzorec: C₃₁H₂₉N₃O₇
MW: 555.59 g/mol
CAS číslo: 216699-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d36883ed84fd30ac7c988f5be9e3a418d8e6169b&vid=4b1164afe8415ba874706fbf10745e08f8686b8c&prid=web&clid=SAPDAM