Tisk…

Sulpho-Cyanine 3 carboxylic acid

Dodavatel: AAT BIOQUEST

260 261
AATB260EA 20520 CZK
AATB260 AATB261
Sulpho-Cyanine 3 carboxylic acid
Sulpho-Cyanine 3 carboxylic acid

A variety of cyanine dyes has been used to label biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.


  • Multiple Bioconjugation Applications
  • Excellent Quality


Upozornení: For research use only.

Vzorec: C₃₁H₃₈N₂O₈S₂
MDL Number: MFCD28978247
CAS číslo: 146368-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caba19f351d1b7c2bfc876abc03ad12d0461c1f6&vid=1b8f296a7b6f51559f9237c40b6f3492273e6153&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR