Tisk…

Coelenterazine n, Ultra Pure Grade calcium indicator

Dodavatel: AAT BIOQUEST
Varování

21155 21161
AATB21155EA 4060 CZK
AATB21155 AATB21161
Coelenterazine n, Ultra Pure Grade calcium indicator
Coelenterazine n

Calcium measurement is critical for numerous biological investigations.


  • Luminescent calcium probe
  • Brighter signal
  • Eliminates autofluorescence

Vzorec: C₃₀H₂₃N₃O₂
CAS číslo: 123437-22-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5468b463abdcd084a0108fc1e78eb1b569057d7d&vid=42382e2990da2bd5330583591195d22c383e4bf9&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR