Coelenterazine f, Ultra Pure Grade calcium indicator
Dodavatel: AAT BIOQUEST
|
Varování
|
21152
21158
AATB21152EA
4060
CZK
AATB21152
AATB21158
Coelenterazine f, Ultra Pure Grade calcium indicator
Coelenterazine f
Calcium measurement is critical for numerous biological investigations.
- Luminescent calcium probe
- Brighter signal
- Recombinant protein
MW:
425,45 g/mol |
CAS číslo:
123437-16-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...