Fluo-8FF™ AM
Dodavatel: AAT BIOQUEST
|
Varování
|
21104
21105
AATB21104EA
8190
CZK
AATB21104
AATB21105
Fluo-8FF™ AM
Fluo-8FF™ AM
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
Upozornení: For research use only.
ADR:
5,II |