Tisk…

Fluo-8L™ potassium salt

Dodavatel: AAT BIOQUEST
Varování

21100 21101
AATB21100EA 8190 CZK
AATB21100 AATB21101
Fluo-8L™ potassium salt
Fluo-8L™ potassium salt

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms


Upozornení: For research use only.

MW: 909,09 g/mol
ADR: 5,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR