Tisk…

Fura Red potassium salt

Dodavatel: AAT BIOQUEST
Varování

21045 21047
AATB21045EA 14330 CZK
AATB21045 AATB21047
Fura Red potassium salt
Fura Red potassium salt

Fura Red is a visible light-excitable fura-2 analog that offers unique possibilities for ratiometric measurement of calcium ion in single cells by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicators.


  • Calcium-dependent fluorescence
  • Ratiometric detection
  • Cell impermeant


Upozornení: For research use only.

MW: 808,98 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1314982b4037a01f941755626e5dfc204dcca3b6&vid=f1770c28d6b8e248d3155749c58940f8ed2e6035&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR