Indo-1 AM ≥95%
Dodavatel: AAT BIOQUEST
|
Varování
|
Synonyms:
1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylester, Indo-1-(acetoxymethyl) ester, 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, Indo-1 AM Ester
21030
21033
AATB21030EA
4060
CZK
AATB21030
AATB21033
Indo-1 AM ≥95%
Indo-1 AM
Calcium measurements are critical for numerous biological investigations.
Vzorec:
C₄₇H₅₁N₃O₂₂ MW: 1009.91 g/mol |
MDL Number:
MFCD00036955 CAS číslo: 112926-02-0 ADR: 5 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...