Fluo-4 Pentapotassium Salt
Dodavatel: AAT BIOQUEST
|
Varování
|
20555
20556
AATB20555EA
12280
CZK
AATB20555
AATB20556
Fluo-4 Pentapotassium Salt
Fluo-4 Pentapotassium Salt
Calcium measurement is critical for numerous biological investigations.
- Calcium-dependent fluorescence
- Steady signal
- Cell impermeant
Upozornení: For research use only.
MW:
927,08 g/mol |
MDL Number:
MFCD03456811 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...