Tisk…

XFD488 NHS Ester, Ultra Pure Grade fluorescent probe

Dodavatel: AAT BIOQUEST
Varování

1810 71901
AATB1810EA 10220 CZK
AATB1810 AATB71901
XFD488 NHS Ester, Ultra Pure Grade fluorescent probe
XFD488 NHS Ester

XFD488 is manufactured by AAT Bioquest, and it has the same chemical structure of Alexa Fluor® 488 (Alexa Fluor® is the trademark of ThermoFisher).


  • pH-insensitive
  • High photostability
  • Brighter signal
  • Stable for 12 months


Quality tested in-house.


Upozornení: For research use only.

ADR: 5,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR