Resorufin alpha-D-glucopyranoside ≥95%
Dodavatel: AAT BIOQUEST
|
Varování
|
14022
14023
AATB14022EA
15640
CZK
AATB14022
AATB14023
Resorufin alpha-D-glucopyranoside ≥95%
Resorufin alpha-D-glucopyranoside
Resorufin alpha-D-glucopyranoside is a sensitive fluorogenic substrate that generates a red fluorescent product (resorufin) upon interaction with alpha-glucosidase.
Vzorec:
C₁₈H₁₇NO₈ MW: 375,33 g/mol |
CAS číslo:
136565-96-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...